Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ald-Ph-PEG6-acid | 2055013-55-1 | 95.3% | 100 MG
SDP

Catalog No. 5000339949
Encompass_Preferred

Ald-Ph-PEG6-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. This compound is a colorless to light yellow viscous liquid.

  • Molecular weight is 485.52
  • Chemical formula is C23H35NO10
  • It appears as a viscous liquid
  • Its color ranges from colorless to light yellow
  • Purity is 95.3%
  • Storage in pure form: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-39949 Supplier Medchemexpress LLC Supplier No. HY130496100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ald-Ph-PEG6-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. This compound is a colorless to light yellow viscous liquid.

  • Molecular weight is 485.52
  • Chemical formula is C23H35NO10
  • It appears as a viscous liquid
  • Its color ranges from colorless to light yellow
  • Purity is 95.3%
  • Storage in pure form: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.