Learn More
Medchemexpress LLC Ald-Ph-PEG6-acid | 2055013-55-1 | 95.3% | 250 MG
SDP
Catalog No. 5000342244
Encompass_Preferred
Ald-Ph-PEG6-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets the protein of interest. PROTACs function by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Composed of two ligands connected by a linker
  • Ligands target an E3 ubiquitin ligase and a protein of interest
  • Functions by exploiting the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-42244 Supplier Medchemexpress LLC Supplier No. 250MGHY130496
Ald-Ph-PEG6-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets the protein of interest. PROTACs function by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Composed of two ligands connected by a linker
  • Ligands target an E3 ubiquitin ligase and a protein of interest
  • Functions by exploiting the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins