Learn More
Medchemexpress LLC Ald-Ph-PEG6-acid | 2055013-55-1 | 95.3% | 50 MG
SDP
Catalog No. 5000340813
Encompass_Preferred
Ald-Ph-PEG6-acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs consist of two distinct ligands connected by a linker; one targets an E3 ubiquitin ligase and the other targets a protein of interest. These molecules work by harnessing the intracellular ubiquitin-proteasome system to selectively degrade target proteins. It appears as a colorless to light yellow viscous liquid.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Targets E3 ubiquitin ligase and a protein of interest
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-40813 Supplier Medchemexpress LLC Supplier No. HY13049650MG
Ald-Ph-PEG6-acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs consist of two distinct ligands connected by a linker; one targets an E3 ubiquitin ligase and the other targets a protein of interest. These molecules work by harnessing the intracellular ubiquitin-proteasome system to selectively degrade target proteins. It appears as a colorless to light yellow viscous liquid.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Targets E3 ubiquitin ligase and a protein of interest
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins