Learn More
Medchemexpress LLC HY-18936 10mg Medchemexpress, Alda-1 CAS:349438-38-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1893610MG
Encompass_Preferred
Medchemexpress, HY-18936 10mg Alda-1 CAS:349438-38-6 Alda-1 is a potent ALDH2 agonist, which activates wild-type ALDH2 and restores near wild-type activity to ALDH2*2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2094