Learn More
Cayman Chemical Aldicarb-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 1795142-83-4, 1 mg
SDP
Catalog No. 502915925
Encompass_Preferred
An internal standard for the quantification of aldicarb by GC- or LC-MS
Catalog No. 50-291-5925 Supplier Cayman Chemical Supplier No. 395801 MG
An internal standard for the quantification of aldicarb by GC- or LC-MS