Learn More
WCM INC Aldex GL15 4-1 Gallon containers per case
Catalog No. NC9934642
Encompass_Preferred
Aldex GL15 case: 4 units of 1 gallon container Aldehyde Management System (liquid)Neutralizes 15 Gallons- 10% Formalin or up to 4% Glutaraldehyde or 3.75% Formaldehydes.Once neutralized safe for drain disposal.
Catalog No. NC9934642 Supplier WCM INC Supplier No. AMSGL15
Aldex GL15 case: 4 units of 1 gallon container Aldehyde Management System (liquid)Neutralizes 15 Gallons- 10% Formalin or up to 4% Glutaraldehyde or 3.75% Formaldehydes.Once neutralized safe for drain disposal.