Learn More
STEMCELL Technologies ALDEFLUOR™ Assay Buffer, Size: 55 mL
SDP
Catalog No. NC9901117
Encompass
Use ALDEFLUOR™ Assay Buffer to optimize the detection of aldehyde dehydrogenase-bright (ALDHbr) cells and maximize fluorescent signal stability during the ALDEFLUOR™ assay.Available either individually or as part of the ALDEFLUOR™ Kit (/products/aldefluor-kit.html), ALDEFLUOR™ Assay Buffer is recommended for use in all procedures performed after ALDH staining to inhibit the active efflux of the ALDEFLUOR™ reaction product and to avoid loss in the assay signal.For detailed information on protocols for using the ALDEFLUOR™ Assay Buffer, please refer to the Product Information Sheet (PIS) (/products/aldefluor-assay-buffer.html#section-protocols-and-documentation).
Catalog No. NC9901117 Supplier STEMCELL Technologies Supplier No. 01702
Use ALDEFLUOR™ Assay Buffer to optimize the detection of aldehyde dehydrogenase-bright (ALDHbr) cells and maximize fluorescent signal stability during the ALDEFLUOR™ assay.Available either individually or as part of the ALDEFLUOR™ Kit (/products/aldefluor-kit.html), ALDEFLUOR™ Assay Buffer is recommended for use in all procedures performed after ALDH staining to inhibit the active efflux of the ALDEFLUOR™ reaction product and to avoid loss in the assay signal.For detailed information on protocols for using the ALDEFLUOR™ Assay Buffer, please refer to the Product Information Sheet (PIS) (/products/aldefluor-assay-buffer.html#section-protocols-and-documentation).