Learn More
STEMCELL Technologies ALDEFLUOR™ DEAB Reagent, 1.5 mM, 1 mL, Size: 1 mL
SDP
Catalog No. NC0435158
Encompass
Use ALDEFLUOR™ DEAB Reagent as a negative control to account for background fluorescence in the ALDEFLUOR™ assay system.Available either individually or as part of the ALDEFLUOR™ Kit (/products/aldefluor-kit.html), this diethylaminobenzaldehyde (DEAB) reagent is an aldehyde dehydrogenase (ALDH) enzyme inhibitor and can be used during the identification of ALDH-bright (ALDHbr) cells.View our additional resources to learn more about the ALDEFLUOR™ reagent system (/products/brands/aldefluor.html).
Catalog No. NC0435158 Supplier STEMCELL Technologies Supplier No. 01705
Use ALDEFLUOR™ DEAB Reagent as a negative control to account for background fluorescence in the ALDEFLUOR™ assay system.Available either individually or as part of the ALDEFLUOR™ Kit (/products/aldefluor-kit.html), this diethylaminobenzaldehyde (DEAB) reagent is an aldehyde dehydrogenase (ALDH) enzyme inhibitor and can be used during the identification of ALDH-bright (ALDHbr) cells.View our additional resources to learn more about the ALDEFLUOR™ reagent system (/products/brands/aldefluor.html).