Learn More
STEMCELL Technologies ALDEFLUOR™ Kit, Size: 1 Kit
SDP
Catalog No. NC9610309
Encompass
ALDEFLUOR™ kit for the identification, evaluation and isolation of stem and progenitor cells expressing high levels of ALDH
Catalog No. NC9610309 Supplier STEMCELL Technologies Supplier No. 01700
ALDEFLUOR™ kit for the identification, evaluation and isolation of stem and progenitor cells expressing high levels of ALDH