Learn More
Medchemexpress LLC Aldehyde-benzyl-PEG5-alkyne | 1378928-83-6 | 97.09% | 336.38 | 100 MG
SDP
Catalog No. 5000348734
Encompass_Preferred
Aldehyde-benzyl-PEG5-alkyne is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules possessing Azide groups. PROTACs themselves consist of two distinct ligands connected by a linker: one ligand targets an E3 ubiquitin ligase, and the other targets a specific protein, leveraging the intracellular ubiquitin-proteasome system for selective protein degradation. This product is intended for research use only.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Contains an alkyne group for CuAAc reactions
  • Utilizes the ubiquitin-proteasome system for selective protein degradation
Catalog No. 50-003-48734 Supplier Medchemexpress LLC Supplier No. HY136140100MG
Aldehyde-benzyl-PEG5-alkyne is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules possessing Azide groups. PROTACs themselves consist of two distinct ligands connected by a linker: one ligand targets an E3 ubiquitin ligase, and the other targets a specific protein, leveraging the intracellular ubiquitin-proteasome system for selective protein degradation. This product is intended for research use only.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Contains an alkyne group for CuAAc reactions
  • Utilizes the ubiquitin-proteasome system for selective protein degradation