Learn More
Medchemexpress LLC Aldehyde-benzyl-PEG5-alkyne | 1378928-83-6 | 97.1% | 336.38 | 50 MG
SDP
Catalog No. 5000351407
Encompass_Preferred
Aldehyde-benzyl-PEG5-alkyne is a PEG-based PROTAC linker. It can be used in the synthesis of PROTACs and acts as a click chemistry reagent. This compound contains an Alkyne group which allows it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. It is intended for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an Alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Azide groups
  • Liquid appearance, colorless to light yellow
  • For research use only
Catalog No. 50-003-51407 Supplier Medchemexpress LLC Supplier No. HY13614050MG
Aldehyde-benzyl-PEG5-alkyne is a PEG-based PROTAC linker. It can be used in the synthesis of PROTACs and acts as a click chemistry reagent. This compound contains an Alkyne group which allows it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. It is intended for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an Alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Azide groups
  • Liquid appearance, colorless to light yellow
  • For research use only