Learn More
Cayman Chemical Aldehyde Dehydrogenase Activity Assay Kit, 96 Well
SDP
Catalog No. 502892340
Encompass_Preferred
Cayman’s Aldehyde Dehydrogenase Assay provides a fluorescence-based method for detecting ALDH activity in tissue homogenates, cell culture samples, and purified ALDH preparations. In the assay, ALDH catalyzes the oxidation of acetaldehyde to acetic acid, along with the concomitant reduction of NAD+ to NADH. NADH reacts with the fluorometric developer to yield a highly fluorescent product which can be analyzed with an excitation wavelength of 530 ± 5 nm and an emission wavelength of 590 ± 5 nm.
Catalog No. 50-289-2340 Supplier Cayman Chemical Supplier No. 70080096 WELL
Cayman’s Aldehyde Dehydrogenase Assay provides a fluorescence-based method for detecting ALDH activity in tissue homogenates, cell culture samples, and purified ALDH preparations. In the assay, ALDH catalyzes the oxidation of acetaldehyde to acetic acid, along with the concomitant reduction of NAD+ to NADH. NADH reacts with the fluorometric developer to yield a highly fluorescent product which can be analyzed with an excitation wavelength of 530 ± 5 nm and an emission wavelength of 590 ± 5 nm.