Learn More
Cayman Chemical Aldehyde Dehydrogenase Polycl
SDP
Catalog No. 502973122
Encompass_Preferred
Immunogen: Human erythrocyte ALDH • Host: Rabbit • Species Reactivity: (+) Human, mouse • Cross Reactivity: (+) ALDH • Applications: WB • MW = 55 kDa
Catalog No. 50-297-3122 Supplier Cayman Chemical Supplier No. 1607201 EA
Immunogen: Human erythrocyte ALDH • Host: Rabbit • Species Reactivity: (+) Human, mouse • Cross Reactivity: (+) ALDH • Applications: WB • MW = 55 kDa