Learn More
Electron Microscopy Sciences Aldehyde-Fuchsin Solution, pH 1.0 500 ML
SDP
Catalog No. 5031755
Encompass_Preferred
Used in Aldehyde-Fuchsin Method (pH 1.0) for stain highly acid sulfated mucosubstances. All mucins except the highly acidic sulfated mucosubstances are aldehyde fuchsin negative at pH values of 1.0 and below. References: Johnson, W.C., Graham, J.H., and Helwig, E.B., J. Invest. Derm., 42:215-224, 1964 and Luna, L.G., (ed), Manual of Histologic Staining Methods of the AFIP, 3rd Ed., McGraw-Hill, c.1968, p. 166
Catalog No. 50-317-55 Supplier Electron Microscopy Sciences Supplier No. 2632801
Used in Aldehyde-Fuchsin Method (pH 1.0) for stain highly acid sulfated mucosubstances. All mucins except the highly acidic sulfated mucosubstances are aldehyde fuchsin negative at pH values of 1.0 and below. References: Johnson, W.C., Graham, J.H., and Helwig, E.B., J. Invest. Derm., 42:215-224, 1964 and Luna, L.G., (ed), Manual of Histologic Staining Methods of the AFIP, 3rd Ed., McGraw-Hill, c.1968, p. 166