Learn More
WCM INC Aldex AMS1010, 2.5 Gallon Aldehyde disposal
Supplier:  WCM INC AMS1010
Encompass_Preferred
Aldex AMS1010, 2.5 Gallon container. Neutralizes and crosslinks used 10% formalin or upto 4% used glutaraldehyde or 3.75% Formaldehyde and converts this waste into a nontoxic, Non-hazardous to be safely disposed in regular trash
Catalog No. NC9128226