Learn More
Assay Technology Aldehyde Monitors, 5/pk, with analysis of one (1) aldehyde included
SDP
Catalog No. NC9911662
Encompass_Preferred
Can be used to monitor for formaldehyde along with other aldehydes. Unlike other aldehyde badges, the 571 can used to monitor formaldehyde in the air for a 15 minutes STEL, as well as a 8 hour PEL, and for indoor air quality, 24 to 72 hours. Aldehydes are collected on a glass fiber filter treated with 2,4-dinitrophenylhydrazine and phosphoric acid. The sample filter is extracted with acetonitrile and analyzed by HPLC using a UV detector. Badges are assembled under a blanket of nitrogen to reduce background levels.
Catalog No. NC9911662 Supplier Assay Technology Supplier No. X571AT
Can be used to monitor for formaldehyde along with other aldehydes. Unlike other aldehyde badges, the 571 can used to monitor formaldehyde in the air for a 15 minutes STEL, as well as a 8 hour PEL, and for indoor air quality, 24 to 72 hours. Aldehydes are collected on a glass fiber filter treated with 2,4-dinitrophenylhydrazine and phosphoric acid. The sample filter is extracted with acetonitrile and analyzed by HPLC using a UV detector. Badges are assembled under a blanket of nitrogen to reduce background levels.