Learn More
Medchemexpress LLC Aldehyde reactive probe (TFA) | 627090-10-2 | 96.4% | 445.41 g/mol | C14H22F3N5O6S | 5 MG
SDP
Catalog No. 5000223371
Encompass_Preferred
Aldehyde reactive probe (TFA) is a biotinylated reagent that reacts with aldehyde groups at apurinic/apyrimidinic (AP) sites to enable detection and quantification of abasic sites in damaged DNA. It is supplied as the trifluoroacetate salt and is used in biochemical assays with streptavidin-based detection.
  • Biotinylated probe for selective labeling of AP sites.
  • Reacts with aldehyde groups generated at abasic sites.
  • Enables streptavidin-based detection and quantification.
  • Suitable for biochemical assays measuring DNA damage and repair.
  • Provided as the trifluoroacetate salt, high purity.
Catalog No. 50-002-23371 Supplier Medchemexpress LLC Supplier No. HY1351365MG
Aldehyde reactive probe (TFA) is a biotinylated reagent that reacts with aldehyde groups at apurinic/apyrimidinic (AP) sites to enable detection and quantification of abasic sites in damaged DNA. It is supplied as the trifluoroacetate salt and is used in biochemical assays with streptavidin-based detection.
  • Biotinylated probe for selective labeling of AP sites.
  • Reacts with aldehyde groups generated at abasic sites.
  • Enables streptavidin-based detection and quantification.
  • Suitable for biochemical assays measuring DNA damage and repair.
  • Provided as the trifluoroacetate salt, high purity.