Learn More
Medchemexpress LLC Aldehyde reactive probe TFA | 627090-10-2 | 96.4% | C14H22F3N5O6S | 10MG
SDP
Catalog No. 5000260528
Encompass_Preferred
Aldehyde reactive probe (TFA) is a biotinylated reagent that selectively reacts with aldehyde groups at apurinic/apyrimidinic (AP) sites in damaged DNA, enabling sensitive detection and quantification using biotin-streptavidin workflows. The trifluoroacetate salt is supplied as a stable solid suitable for labeling, analytical assays, and downstream detection protocols.
  • Enables selective labeling of AP sites in DNA.
  • Facilitates detection and quantification via biotin-streptavidin systems.
  • High purity suitable for analytical applications.
  • Available in multiple pack sizes for assay scale flexibility.
  • Trifluoroacetate salt form improves solubility and handling.
Catalog No. 50-002-60528 Supplier Medchemexpress LLC Supplier No. HY13513610MG
Aldehyde reactive probe (TFA) is a biotinylated reagent that selectively reacts with aldehyde groups at apurinic/apyrimidinic (AP) sites in damaged DNA, enabling sensitive detection and quantification using biotin-streptavidin workflows. The trifluoroacetate salt is supplied as a stable solid suitable for labeling, analytical assays, and downstream detection protocols.
  • Enables selective labeling of AP sites in DNA.
  • Facilitates detection and quantification via biotin-streptavidin systems.
  • High purity suitable for analytical applications.
  • Available in multiple pack sizes for assay scale flexibility.
  • Trifluoroacetate salt form improves solubility and handling.