Learn More
Cayman Chemical Aldehyde Reactive Probe (trifluoroacetate salt), Purity: ≥95%, CAS: 627090-10-2, 25 mg
SDP
Catalog No. 502889076
Encompass_Preferred
A biotinylated reagent used for the detection and quantification of apurinic/apyrimidinic (AP) sites in damaged DNA; reacts with aldehyde groups formed when reactive oxygen species depurinate DNA, thereby covalently linking biotin to these AP sites.
Catalog No. 50-288-9076 Supplier Cayman Chemical Supplier No. 1000935025 MG
A biotinylated reagent used for the detection and quantification of apurinic/apyrimidinic (AP) sites in damaged DNA; reacts with aldehyde groups formed when reactive oxygen species depurinate DNA, thereby covalently linking biotin to these AP sites.