Learn More
Cayman Chemical Aldehyde Reactive Probe trif
SDP
Supplier:  Cayman Chemical 1000935050 MG
Encompass_Preferred
A biotinylated reagent used for the detection and quantification of apurinic/apyrimidinic (AP) sites in damaged DNA; reacts with aldehyde groups formed when reactive oxygen species depurinate DNA, thereby covalently linking biotin to these AP sites.
Catalog No. 50-288-9443