Learn More
Bioworld ALDEHYDE SEPAROPOREAGAROS
Supplier:  Bioworld 201810582
Encompass
ALDEHYDE SEPAROPORE (AGAROSE) 4B-CL: Specifications: Active group: -CHO; Particle size (range): 50-175m; Molecular Weight range: 6x104 - 2x107; Matrix: Separopore 4B-CL (Crosslinked Agarose 4% Beads); pH stability: 4-9; Flow specificatio...
Catalog No. 50-197-812