Learn More
Electron Microscopy Sciences Aldehyde/Silane Slide With ES & Bar Code
SDP
Catalog No. 5031442
Encompass_Preferred
Offers a uniform coating of aldehydes for attaching amine-modified DNA. ES coating produces uniform spot size.
Catalog No. 50-314-42 Supplier Electron Microscopy Sciences Supplier No. 63476EB
Offers a uniform coating of aldehydes for attaching amine-modified DNA. ES coating produces uniform spot size.