Learn More
WCM INC Aldex825, 55 Gallon Drum, Treats 825 Gallons of Aldehyde Waste
Catalog No. NC3781045
Encompass_Preferred
Aldex 825, 55 Gallon drum. Neutralizer converts liquid aldehyde waste (10% Formalin, 4% Glutaraldehydes or 3.75% Formaldehydes) into a non hazardous liquid safe for drain disposal. Drum treats a total of 825 Gallons of Aldehyde waste.
Catalog No. NC3781045 Supplier WCM INC Supplier No. AMSGL825
Aldex 825, 55 Gallon drum. Neutralizer converts liquid aldehyde waste (10% Formalin, 4% Glutaraldehydes or 3.75% Formaldehydes) into a non hazardous liquid safe for drain disposal. Drum treats a total of 825 Gallons of Aldehyde waste.