Learn More
WCM INC Aldex GL1 8oz 12/PK, 8 oz treats 1G, total 12 Gallons
Catalog No. NC9671456
Encompass_Preferred
Aldex GL1, 8oz bottles, 12 bottles total. 8 oz liquid treats 1 Gallon. Neutralizer converts liquid aldehyde waste (10% Formalin, 4% Glutaraldehydes or 3.75% Formaldehydes) into a non hazardous liquid safe for drain disposal. Treats a total of 12 Gallons
Catalog No. NC9671456 Supplier WCM INC Supplier No. AMSGL112
Aldex GL1, 8oz bottles, 12 bottles total. 8 oz liquid treats 1 Gallon. Neutralizer converts liquid aldehyde waste (10% Formalin, 4% Glutaraldehydes or 3.75% Formaldehydes) into a non hazardous liquid safe for drain disposal. Treats a total of 12 Gallons