Learn More
Sino Biological Anti-ALDH1A1 Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 11388MM03P25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ALDH1A1 / ALDH1 (rh ALDH1A1 / ALDH1; Catalog#11388-H07E; NP_000680.2; Ser2-Ser501) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-228-3621