Learn More
Medchemexpress LLC HY-P7474 10ug Medchemexpress, ALDH1A1, His, Human CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP747410UG
Encompass_Preferred
Medchemexpress, HY-P7474 10ug ALDH1A1, His, Human CAS: Recombinant Human Aldehyde Dehydrogenase 1-A1, His is a human recombinant ALDH1A1 with an N-terminal Met and His tag produced in E. coli. Recombinant Human Aldehyde Dehydrogenase 1-A1, His contains 501 amino acids (1-501 a.a.). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3169