Learn More
Bioss ALDH1A1 - N-terminal region An
SDP
Catalog No. 5000064741
Encompass_Preferred
ALDH1A1 (N-terminal region) Primary Antibody Unconjugated Host Species Mouse Target Species Human Clonality Monoclonal Applications WB M562Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol
Catalog No. 50-000-64741 Supplier Bioss Supplier No. BSM70333M
ALDH1A1 (N-terminal region) Primary Antibody Unconjugated Host Species Mouse Target Species Human Clonality Monoclonal Applications WB M562Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol