Learn More
Aviva Systems Biology Corp ALDH1L1 Antibody - middle region (ARP52331_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52331_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ALDH1L1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ALDH1L1. Alternate names: DKFZp781N0997, FTHFD, FDH, 10-fTHFApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Yeast, Zebrafish
Catalog No. 50-014-60714