Learn More
Aviva Systems Biology Corp ALDH3A1 Antibody - C-terminal region (ARP59747_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59747_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ALDH3A1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human ALDH3A1. Alternate names: ALDH3, ALDHIII, MGC10406ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-286-805