Learn More
Aviva Systems Biology Corp ALDH3A1 Antibody - N-terminal region (ARP59748_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59748_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ALDH3A1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ALDH3A1. Alternate names: ALDH3, ALDHIII, MGC10406ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Rat
Catalog No. 50-286-806