Learn More
Sino Biological ALDH3A1 Antibody, Rabbit MAb 100uL
Supplier:  Sino Biological 12523R316100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human ALDH3A1 (rh ALDH3A1; Catalog#12523-H07B; AAH04370.1; Met1-His453).
Catalog No. 50-224-7908