Learn More
Sino Biological ALDH3A1 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 12523T60100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ALDH3A1 (rh ALDH3A1; Catalog#12523-H07B; AAH04370.1; Met1-His453). ALDH3A1 specific IgG was purified by Human ALDH3A1 affinity chromatography.
Catalog No. 50-224-8689