Learn More
Sino Biological ALDH3A1 Antibody, Rabbit PAb 100uL
Supplier:  Sino Biological 12523RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ALDH3A1 (rh ALDH3A1; Catalog#12523-H07B; AAH04370.1; Met1-His453). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-224-7959