Learn More
Aviva Systems Biology Corp ALDH3A2 Antibody - C-terminal region (ARP44274_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44274_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ALDH3A2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human ALDH3A2. Alternate names: ALDH10, DKFZp686E23276, FALDH, SLSApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58766