Learn More
Aviva Systems Biology Corp ALDH3B1 Antibody - N-terminal region (ARP54347_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54347_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ALDH3B1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ALDH3B1. Alternate names: ALDH4, ALDH7, FLJ26433ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-61182