Learn More
Aviva Systems Biology Corp ALDH4A1 Antibody - N-terminal region (ARP45696_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45696_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ALDH4A1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human ALDH4A1. Alternate names: RP11-128M10.1, ALDH4, P5CD, P5CDh, P5CDhL, P5CDhSApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-59104