Learn More
ABclonal Technology ALDH7A1 Rabbit mAb
SDP
Catalog No. 5000008101
Encompass_Preferred
The protein encoded by this gene is a member of subfamily 7 in the aldehyde dehydrogenase gene family These enzymes are thought to play a major role in the detoxification of aldehydes generated by alcohol metabolism and lipid peroxidation This particular member has homology to a previously described protein from the green garden pea the 26g pea turgor protein It is also involved in lysine catabolism that is known to occur in the mitochondrial matrix Recent reports show that this protein is found both in the cytosol and the mitochondria and the two forms likely arise from the use of alternative translation initiation sites An additional variant encoding a different isoform has also been found for this gene Mutations in this gene are associated with pyridoxine-dependent epilepsy Several related pseudogenes have also been identified
Catalog No. 50-000-08101 Supplier ABclonal Technology Supplier No. A20888100UL
The protein encoded by this gene is a member of subfamily 7 in the aldehyde dehydrogenase gene family These enzymes are thought to play a major role in the detoxification of aldehydes generated by alcohol metabolism and lipid peroxidation This particular member has homology to a previously described protein from the green garden pea the 26g pea turgor protein It is also involved in lysine catabolism that is known to occur in the mitochondrial matrix Recent reports show that this protein is found both in the cytosol and the mitochondria and the two forms likely arise from the use of alternative translation initiation sites An additional variant encoding a different isoform has also been found for this gene Mutations in this gene are associated with pyridoxine-dependent epilepsy Several related pseudogenes have also been identified