Learn More
Aviva Systems Biology Corp ALDH9A1 Antibody - C-terminal region (ARP59590_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59590_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ALDH9A1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human ALDH9A1. Alternate names: ALDH4, ALDH7, ALDH9, E3, TMABADHApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-752