Learn More
Cayman Chemical AldIcarb sulfon 250mg
SDP
Catalog No. 502935947
Encompass_Preferred
A metabolite and degradation product of aldicarb; reduces the number of male T. semipenetrans second-stage larvae that reach the third, fourth, and adult stages of development at 100 μg/ml
Catalog No. 50-293-5947 Supplier Cayman Chemical Supplier No. 24210250 MG
Add to Cart
Add to Cart
A metabolite and degradation product of aldicarb; reduces the number of male T. semipenetrans second-stage larvae that reach the third, fourth, and adult stages of development at 100 μg/ml