Learn More
Cayman Chemical Aldicarb sulfone, Purity: ≥95%, CAS: 1646-88-4, 50 mg
SDP
Catalog No. 502935482
Encompass_Preferred
A metabolite and degradation product of aldicarb; reduces the number of male T. semipenetrans second-stage larvae that reach the third, fourth, and adult stages of development at 100 μg/ml
Catalog No. 50-293-5482 Supplier Cayman Chemical Supplier No. 2421050 MG
A metabolite and degradation product of aldicarb; reduces the number of male T. semipenetrans second-stage larvae that reach the third, fourth, and adult stages of development at 100 μg/ml