Learn More
Medchemexpress LLC Aldicarb (sulfone) | 1646-88-4 | 99.46% | 222.26 | 50 MG
SDP
Catalog No. 5000386295
Encompass_Preferred
Aldicarb sulfone is a metabolite of the carbamate pesticide Aldicarb, exhibiting insecticidal activity. It is also known to be a cholinesterase inhibitor.
  • Inhibits egg hatching in *Tylenchulus semipenetrans*
  • Reduces nematode motility
  • Hindered male larval development
  • Decreased plasma pseudocholinesterase levels in rats
  • Decreased red blood cell cholinesterase levels in rats
Catalog No. 50-003-86295 Supplier Medchemexpress LLC Supplier No. HY1753050MG
Aldicarb sulfone is a metabolite of the carbamate pesticide Aldicarb, exhibiting insecticidal activity. It is also known to be a cholinesterase inhibitor.
  • Inhibits egg hatching in *Tylenchulus semipenetrans*
  • Reduces nematode motility
  • Hindered male larval development
  • Decreased plasma pseudocholinesterase levels in rats
  • Decreased red blood cell cholinesterase levels in rats