Learn More
Cayman Chemical Aldicarb Sulfoxide, Purity: ≥98%, CAS: 1646-87-3, 25 mg
SDP
Catalog No. 502935643
Encompass_Preferred
A metabolite and degradation product of aldicarb; reduces the number of male T. semipenetrans second-stage larvae that reach the third, fourth, and adult stages of development at 10 μg/ml; inhibits carboxylesterase and cholinesterase in zebrafish (IC50 = 10 μM for both)
Catalog No. 50-293-5643 Supplier Cayman Chemical Supplier No. 2421125 MG
A metabolite and degradation product of aldicarb; reduces the number of male T. semipenetrans second-stage larvae that reach the third, fourth, and adult stages of development at 10 μg/ml; inhibits carboxylesterase and cholinesterase in zebrafish (IC50 = 10 μM for both)