Learn More
Enzo Life Sciences Aldolase A monoclonal antibody (AT3F9) (100µl)
SDP
Supplier:  Enzo Life Sciences ENZABS7600100
Encompass_Preferred
Clone: AT3F9. Isotype: IgG2aκ. Immunogen: Recombinant human Aldolase A (aa 1-364) purified from E. coli. Species reactivity: Human. Applications: ELISA, Flow Cytometry, ICC, IF, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.4, containing 0.02% sodium azide and 10% glycerol.
Catalog No. 50-201-3299