Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aldolase, Rabbit muscle | 9024-52-6 | 500 U
SDP

Catalog No. 5000400332
Encompass_Preferred

Aldolase, Rabbit muscle is a glycolytic enzyme and a component of the VATPase complex. It causes fructose 1, 6-diphosphate to decompose into dihydroxyacetone and glyceraldehyde 3-phosphate. This product is for research use only.

  • Liquid appearance
  • White to off-white color
  • EC Number: 4.1.2.13
  • Specific activity of ≥10 U/mg protein
  • Unit defined by an increase of 1.0 at A240 per minute at 25°C, pH 7.5 with the hydrazine/3-phosphoglyceraldehyde assay
  • Classified as an enzyme and biochemical detection enzyme
  • Relevant to metabolic and endocrine disease research
  • Identified as an endogenous metabolite

Catalog No. 50-004-00332 Supplier Medchemexpress LLC Supplier No. HYP2726500U
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aldolase, Rabbit muscle is a glycolytic enzyme and a component of the VATPase complex. It causes fructose 1, 6-diphosphate to decompose into dihydroxyacetone and glyceraldehyde 3-phosphate. This product is for research use only.

  • Liquid appearance
  • White to off-white color
  • EC Number: 4.1.2.13
  • Specific activity of ≥10 U/mg protein
  • Unit defined by an increase of 1.0 at A240 per minute at 25°C, pH 7.5 with the hydrazine/3-phosphoglyceraldehyde assay
  • Classified as an enzyme and biochemical detection enzyme
  • Relevant to metabolic and endocrine disease research
  • Identified as an endogenous metabolite

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.