Learn More
Medchemexpress LLC Aldometanib | 2904601-67-6 | 99.7% | 593.45 | 50 MG
SDP
Catalog No. 5000361335
Encompass_Preferred
Aldometanib is an orally active aldolase inhibitor that activates lysosomal adenosine monophosphate-activated protein kinase (AMPK) and reduces blood glucose. This compound is suitable for research into metabolic homeostasis.
  • Activates lysosomal AMPK by generating a pseudo-starvation signal.
  • Decreases fasting blood glucose and improves glucose tolerance.
  • Alleviates fatty liver and nonalcoholic steatohepatitis.
  • Extends lifespan and rejuvenates muscle function.
This item is not returnable. View return policy
Catalog No. 50-003-61335 Supplier Medchemexpress LLC Supplier No. HY14818950MG
Aldometanib is an orally active aldolase inhibitor that activates lysosomal adenosine monophosphate-activated protein kinase (AMPK) and reduces blood glucose. This compound is suitable for research into metabolic homeostasis.
  • Activates lysosomal AMPK by generating a pseudo-starvation signal.
  • Decreases fasting blood glucose and improves glucose tolerance.
  • Alleviates fatty liver and nonalcoholic steatohepatitis.
  • Extends lifespan and rejuvenates muscle function.