Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Blood Typing Anti-Sera, Anti-A, Anti-B and Anti Rh
SCI_ED

Catalog No. s07803 Shop All Aldon Products
Change view
Click to view available options
Includes:
Anti-a and anti-b
Anti-rh
Type a, b and rh
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S07803 Type a, b and rh
S07802 Anti-a and anti-b
S07804 Anti-rh
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S07803 Supplier Aldon Corporation Supplier No. IS3171
Only null left
Add to Cart
Add to Cart

Use for blood typing of real human blood. Each vial contains enough antisera for 75 students. Refrigeration recommended until used. Materials from human-derived cell lines were found to be negative in tests for Hepatitis B, HIV and EBV.

  • Available in 5mL vials that contain enough antisera for 75 students
  • Freeze-dried samples; can be kept without refrigeration
  • Shows negative results for Hepatitis B, HIV and EBV when tested with human-derived cell lines

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Use for blood typing of real human blood
Includes Type a, b and rh
Quantity 1
Dimensions (L x W x H) 3 x .0.5 x 0.5 in.
Class Size 75 Students
Product Type Blood Typing Anti-Sera
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.