Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Electrophoresis and DNA Fragment Length Determination Kit
GREENER_CHOICESCI_ED

Catalog No. S06973 Shop All Aldon Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S06973 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S06973 Supplier Innovating Science™ Supplier No. IS3300
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Room temperature DNA (Enough to run 10 gels)
  • Prepared agarose
  • TBE electrophoresis buffer
  • DNA stain

Requires

Agarose electrophoresis chambers, power supplies and micropipettes capable of measuring 10μL

Ideal for students to learn about the process and techniques associated agarose electrophoresis.

  • Collect water samples and use a low range pH indicator with a colorimetric comparison chart.
  • Kit Includes:
    • 15 mL low range pH indicator
    • 6 test tubes
    • Color chart

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) For students to learn about the process and techniques associated agarose electrophoresis
Includes 200 mL 0.8% agarose, 500 mL 5x tris borate eta buffer, 60 mL 20x dna stain, lambda dna ecori digest with loading dye, lambda dna hindiii digest with loading dye, lambda dna ecori/hind iii digest with loading dye
Quantity 1
Dimensions (L x W x H) 9.75 x 6.25 x 4 in.
Class Size 10 Lab Groups
Product Type Electrophoresis and DNA Fragment Length Determination Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.