Learn More
Cayman Chemical Aldosteron 10mg
SDP
Catalog No. 502970203
Encompass_Preferred
A steroid hormone secreted by the adrenal cortex that controls sodium and potassium balance
Catalog No. 50-297-0203 Supplier Cayman Chemical Supplier No. 1527310 MG
A steroid hormone secreted by the adrenal cortex that controls sodium and potassium balance