Learn More
Cayman Chemical Aldosteron 21sulfATsodIu 10mg
SDP
Catalog No. 502895505
Encompass_Preferred
A metabolite of aldosterone; has weak mineralocorticoid activity that is <1% that of aldosterone in rats at 10-100 UG
Catalog No. 50-289-5505 Supplier Cayman Chemical Supplier No. 900345010 MG
A metabolite of aldosterone; has weak mineralocorticoid activity that is <1% that of aldosterone in rats at 10-100 UG