Learn More
Eurofins Discoverx Aldosterone, 5 mg. (92-1131)
Catalog No. 501999664
Encompass
Aldosterone is an adrenal steroid hormone that plays an important role in blood pressure regulation. Acts on the nephron to increase reabsorption of ions and water in the kidney and increase blood pressure and volume. Controls amount of sodium and potassium in the bloodstream. Provided as 5 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9664 Supplier Eurofins Discoverx Supplier No. 921131
Aldosterone is an adrenal steroid hormone that plays an important role in blood pressure regulation. Acts on the nephron to increase reabsorption of ions and water in the kidney and increase blood pressure and volume. Controls amount of sodium and potassium in the bloodstream. Provided as 5 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.